COMH04236B96 Combi-Blocks, Inc.

[1-(1,3-Benzodioxol-5-ylmethyl)-2,5-dioxoimidazolidin-4-yl]acetic acid

  • MDL number MFCD28142973
  • Cas Number : 1638709-80-4
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8686
Availability details for
Quantity
  • Availability
  • Add to Cart