COMH04236BA3 Combi-Blocks, Inc.

6-(Chloromethyl)-n,n-dimethyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00463462
  • Cas Number : 21320-37-6
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8699
Availability details for
Quantity
  • Availability
  • Add to Cart