COMH04236BC3 Combi-Blocks, Inc.

(2-(2-[2-(2-Aminoethyl)-4,5-dimethoxybenzyl]-4,5-dimethoxyphenyl)ethyl)amine dihydrochloride

  • MDL number MFCD27959820
  • Cas Number : 1328085-12-6
  • Molecular Weight : 447.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8731
Availability details for
Quantity
  • Availability
  • Add to Cart