COMH04236BC6 Combi-Blocks, Inc.

1-[(4-Fluorophenyl)sulfonyl]piperidine-4-carbohydrazide

  • MDL number MFCD01821806
  • Cas Number : 312534-13-7
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8734
Availability details for
Quantity
  • Availability
  • Add to Cart