COMH04236BD1 Combi-Blocks, Inc.

2-[(4-Allyl-5-([(4-methylphenyl)amino]methyl)-4h-1,2,4-triazol-3-yl)thio]acetohydrazide

  • MDL number MFCD02255646
  • Cas Number : 1071368-63-2
  • Molecular Weight : 332.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8745
Availability details for
Quantity
  • Availability
  • Add to Cart