COMH04236BD6 Combi-Blocks, Inc.

2-Hydrazino-4-(piperidin-1-ylcarbonyl)pyridine

  • MDL number MFCD16726566
  • Cas Number : 1247561-58-5
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8750
Availability details for
Quantity
  • Availability
  • Add to Cart