COMH04236BF9 Combi-Blocks, Inc.

3-[1-Methyl-3-(4-methylphenyl)-1h-pyrazol-4-yl]propanoic acid

  • MDL number MFCD11900103
  • Cas Number : 1216073-28-7
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8786
Availability details for
Quantity
  • Availability
  • Add to Cart