COMH04236C04 Combi-Blocks, Inc.

tert-Butyl n-(1-carbamoyl-2-methylpropyl)carbamate

  • MDL number MFCD24390407
  • Cas Number : 73674-46-1
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8797
Availability details for
Quantity
  • Availability
  • Add to Cart