COMH04236C34 Combi-Blocks, Inc.

7-Methyl-2-pyrrolidin-1-yl[1,2,4]triazolo[1,5-a]pyrimidin-5(4h)-one

  • MDL number MFCD27936942
  • Cas Number : 1428139-18-7
  • Molecular Weight : 219.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8845
Availability details for
Quantity
  • Availability
  • Add to Cart