COMH04236C3F Combi-Blocks, Inc.

2-Bromo-n-(4-methoxyphenyl)butanamide

  • MDL number MFCD02939927
  • Cas Number : 188491-09-0
  • Molecular Weight : 272.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8856
Availability details for
Quantity
  • Availability
  • Add to Cart