COMH04236C50 Combi-Blocks, Inc.

4-[((5-[(4-Chlorophenoxy)methyl]-1,3,4-thiadiazol-2-yl)carbonyl)amino]benzoic acid

  • MDL number MFCD12028252
  • Cas Number : 1142202-91-2
  • Molecular Weight : 389.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8873
Availability details for
Quantity
  • Availability
  • Add to Cart