COMH04236C65 Combi-Blocks, Inc.

Methyl 6-(4-methoxyphenyl)-2-oxo-1,2-dihydropyridine-3-carboxylate

  • MDL number MFCD19103585
  • Cas Number : 125031-50-7
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8894
Availability details for
Quantity
  • Availability
  • Add to Cart