COMH04236C73 Combi-Blocks, Inc.

2-([(3,5-Dimethylphenyl)amino]methyl)-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3-dione

  • MDL number MFCD17069386
  • Cas Number : 1241674-88-3
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8908
Availability details for
Quantity
  • Availability
  • Add to Cart