COMH04236C7D Combi-Blocks, Inc.

2-Cyclopentyl-5-methyl-1h-benzimidazole

  • MDL number MFCD18807579
  • Cas Number : 221548-36-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8918
Availability details for
Quantity
  • Availability
  • Add to Cart