COMH04236CC8 Combi-Blocks, Inc.

2-(2-Chlorobenzyl)-1h-benzimidazole

  • MDL number MFCD09880336
  • Cas Number : 213133-88-1
  • Molecular Weight : 242.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8993
Availability details for
Quantity
  • Availability
  • Add to Cart