COMH04236CFE Combi-Blocks, Inc.

1-(4-Bromo-3,5-dimethyl-1h-pyrazol-1-yl)propan-2-ol

  • MDL number MFCD16686973
  • Cas Number : 1249695-94-0
  • Molecular Weight : 233.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9047
Availability details for
Quantity
  • Availability
  • Add to Cart