COMH04236D02 Combi-Blocks, Inc.

Ethyl 5-(difluoromethyl)-1-methyl-1h-pyrazole-3-carboxylate

  • MDL number MFCD24849134
  • Cas Number : 1437312-22-5
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9051
Availability details for
Quantity
  • Availability
  • Add to Cart