COMH04236D39 Combi-Blocks, Inc.

N-(5-Acetyl-4-methyl-1,3-thiazol-2-yl)-4-chlorobenzamide

  • MDL number MFCD01930865
  • Cas Number : 303127-33-5
  • Molecular Weight : 294.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9106
Availability details for
Quantity
  • Availability
  • Add to Cart