COMH04236D5C Combi-Blocks, Inc.

5-Chloro-2-[(2,6-dichlorobenzyl)oxy]benzoyl chloride

  • MDL number MFCD12198010
  • Cas Number : 1160260-26-3
  • Molecular Weight : 350.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9141
Availability details for
Quantity
  • Availability
  • Add to Cart