COMH04236D66 Combi-Blocks, Inc.

(4-Bromophenoxy)acetyl chloride

  • MDL number MFCD02295745
  • Cas Number : 16738-06-0
  • Molecular Weight : 249.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9151
Availability details for
Quantity
  • Availability
  • Add to Cart