COMH04236D6F Combi-Blocks, Inc.

4-Amino-5-(3,5-dichlorophenyl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD00817006
  • Cas Number : 144729-37-3
  • Molecular Weight : 261.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9160
Availability details for
Quantity
  • Availability
  • Add to Cart