COMH04236D72 Combi-Blocks, Inc.

3-(4-Nitrobenzyl)-1,3-benzoxazol-2(3h)-one

  • MDL number MFCD00692457
  • Cas Number : 313986-04-8
  • Molecular Weight : 270.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9163
Availability details for
Quantity
  • Availability
  • Add to Cart