COMH04236D74 Combi-Blocks, Inc.

(2Z)-1-Cyclopropyl-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD04969911
  • Cas Number : 21666-68-2
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9165
Availability details for
Quantity
  • Availability
  • Add to Cart