COMH04236D86 Combi-Blocks, Inc.

3-Cyclopropyl-5-(difluoromethyl)-1h-pyrazole

  • MDL number MFCD08457676
  • Cas Number : 1042768-00-2
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9183
Availability details for
Quantity
  • Availability
  • Add to Cart