COMH04236D8A Combi-Blocks, Inc.

Ethyl 3-tert-butyl-1-phenyl-1h-pyrazole-5-carboxylate

  • MDL number MFCD13248664
  • Cas Number : 1199215-68-3
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9187
Availability details for
Quantity
  • Availability
  • Add to Cart