COMH04236D9D Combi-Blocks, Inc.

[1-(Pyridin-2-ylmethyl)piperidin-3-yl]methanol dihydrochloride

  • MDL number MFCD13857471
  • Cas Number : 1332531-03-9
  • Molecular Weight : 279.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9206
Availability details for
Quantity
  • Availability
  • Add to Cart