COMH04236DA3 Combi-Blocks, Inc.

5-tert-Butyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD04969091
  • Cas Number : 126231-21-8
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9212
Availability details for
Quantity
  • Availability
  • Add to Cart