COMH04236DCE Combi-Blocks, Inc.

2,9-Dimethyl-3,5-diphenyl-7h-furo[3,2-g]chromen-7-one hydrazone

  • MDL number MFCD13969077
  • Cas Number : 1092333-69-1
  • Molecular Weight : 380.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9255
Availability details for
Quantity
  • Availability
  • Add to Cart