COMH04236DCF Combi-Blocks, Inc.

Butyl 3-aminobenzoate

  • MDL number MFCD01112170
  • Cas Number : 26218-03-1
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9256
Availability details for
Quantity
  • Availability
  • Add to Cart