COMH04236DD2 Combi-Blocks, Inc.

1-Bromo-4-(sulfinylamino)benzene

  • MDL number MFCD03249681
  • Cas Number : 26516-62-1
  • Molecular Weight : 218.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9259

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9259
Availability details for
Quantity
  • Availability
  • Add to Cart