COMH04236DD4 Combi-Blocks, Inc.

[3-(1-Methyl-1h-imidazol-2-yl)phenyl]amine dihydrochloride

  • MDL number MFCD14584109
  • Cas Number : 1401425-69-1
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9261
Availability details for
Quantity
  • Availability
  • Add to Cart