COMH04236DD6 Combi-Blocks, Inc.

3-(Pentylsulfanyl)-1h-1,2,4-triazol-5-amine

  • MDL number MFCD01420311
  • Cas Number : 312290-44-1
  • Molecular Weight : 186.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9263
Availability details for
Quantity
  • Availability
  • Add to Cart