COMH04236DE1 Combi-Blocks, Inc.

2-(4-Bromophenyl)-6-chloroquinoline-4-carbonyl chloride

  • MDL number MFCD03421299
  • Cas Number : 1160263-28-4
  • Molecular Weight : 381.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9274
Availability details for
Quantity
  • Availability
  • Add to Cart