COMH04236DE9 Combi-Blocks, Inc.

7,8-Dimethyl-2-(4-methylphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD12198066
  • Cas Number : 1160261-34-6
  • Molecular Weight : 309.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9282
Availability details for
Quantity
  • Availability
  • Add to Cart