COMH04236DEE Combi-Blocks, Inc.

2-[(4-Methyl-5-phenyl-4h-1,2,4-triazol-3-yl)thio]acetohydrazide

  • MDL number MFCD02679122
  • Cas Number : 120873-48-5
  • Molecular Weight : 263.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9287
Availability details for
Quantity
  • Availability
  • Add to Cart