COMH04236E27 Combi-Blocks, Inc.

Ethyl (2z)-2-cyano-3-(2-furyl)acrylate

  • MDL number MFCD00100358
  • Cas Number : 23973-22-0
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9344
Availability details for
Quantity
  • Availability
  • Add to Cart