COMH04236E3C Combi-Blocks, Inc.

3-[4-(4-Chlorophenyl)piperazin-1-yl]-4-methoxycyclobut-3-ene-1,2-dione

  • MDL number MFCD28954482
  • Cas Number : 1858257-22-3
  • Molecular Weight : 306.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9365
Availability details for
Quantity
  • Availability
  • Add to Cart