COMH04236E45 Combi-Blocks, Inc.

[(1-Ethylpropyl)amino](oxo)acetic acid

  • MDL number MFCD12028232
  • Cas Number : 1142202-55-8
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9374
Availability details for
Quantity
  • Availability
  • Add to Cart