COMH04236E74 Combi-Blocks, Inc.

N-(2-Furylmethyl)-n-[(5-methoxy-1h-indol-3-yl)methyl]amine

  • MDL number MFCD11982921
  • Cas Number : 1114597-54-4
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9421
Availability details for
Quantity
  • Availability
  • Add to Cart