COMH04236E9B Combi-Blocks, Inc.

1-(Methylsulfonyl)-5-nitroindoline

  • MDL number MFCD01571905
  • Cas Number : 312523-56-1
  • Molecular Weight : 242.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9460
Availability details for
Quantity
  • Availability
  • Add to Cart