COMH04236EEE Combi-Blocks, Inc.

2-Methyl-6-pyrrolidin-3-ylpyridin-4-ol hydrochloride

  • MDL number MFCD27756607
  • Cas Number : 1381126-87-9
  • Molecular Weight : 214.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9543
Availability details for
Quantity
  • Availability
  • Add to Cart