COMH04236F02 Combi-Blocks, Inc.

3-Phenyl-2h-1,4-benzothiazine-2-carboxylic acid

  • MDL number MFCD12197731
  • Cas Number : 1160264-28-7
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9563
Availability details for
Quantity
  • Availability
  • Add to Cart