COMH04236F1A Combi-Blocks, Inc.

4-[(3,4-Dichlorobenzyl)oxy]benzoyl chloride

  • MDL number MFCD12197980
  • Cas Number : 1160251-27-3
  • Molecular Weight : 315.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9587
Availability details for
Quantity
  • Availability
  • Add to Cart