COMH04236F23 Combi-Blocks, Inc.

5'-Amino-2,3'-bithiophene-4'-carboxylic acid

  • MDL number MFCD21908958
  • Cas Number : 1368920-44-8
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9596
Availability details for
Quantity
  • Availability
  • Add to Cart