COMH04236F2D Combi-Blocks, Inc.

3-(Pyridin-2-yl)propanoic acid hydrochloride

  • MDL number MFCD11696415
  • Cas Number : 15197-75-8
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9606
Availability details for
Quantity
  • Availability
  • Add to Cart