COMH04236F4F Combi-Blocks, Inc.

2-Amino-n-[3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD00423371
  • Cas Number : 20878-52-8
  • Molecular Weight : 280.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9640
Availability details for
Quantity
  • Availability
  • Add to Cart