COMH04236F53 Combi-Blocks, Inc.

(1-Ethyl-1h-indol-5-yl)methanol

  • MDL number MFCD12026987
  • Cas Number : 1030426-01-7
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9644
Availability details for
Quantity
  • Availability
  • Add to Cart