COMH04236F56 Combi-Blocks, Inc.

1-(5-Bromothien-2-yl)butan-1-one

  • MDL number MFCD06657522
  • Cas Number : 106652-43-1
  • Molecular Weight : 233.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9647
Availability details for
Quantity
  • Availability
  • Add to Cart