COMH04236F62 Combi-Blocks, Inc.

2-(3-Hydroxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD01459896
  • Cas Number : 32366-58-8
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9659
Availability details for
Quantity
  • Availability
  • Add to Cart