COMH04236F6C Combi-Blocks, Inc.

2-Bromo-n-(2,5-dimethoxyphenyl)propanamide

  • MDL number MFCD01311977
  • Cas Number : 3245-91-8
  • Molecular Weight : 288.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-9669
Availability details for
Quantity
  • Availability
  • Add to Cart